Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
REV
Entrez Gene ID
836190
Gene Name
Homeobox-leucine zipper family protein / lipid-binding START domain-containing protein
Gene Aliases
AT5G60690, HOMEODOMAIN-LEUCINE ZIPPER PROTEIN, HOMEODOMAIN-LEUCINE ZIPPER PROTEIN IFL1, IFL, IFL1, INTERFASCICULAR FIBERLESS, INTERFASCICULAR FIBERLESS 1, MUP24.16, MUP24_16, REVOLUTA
Chromosome Location
Chr.5: 24396981 - 24402195 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_125462.4 NP_200877.1 11 - 12 2118 70
GenBank mRNA AF188994.1 - 11 - 12 1869 70
AF233592.1 - 11 - 12 1552 70
AK229564.1 - 11 - 12 2076 70

View Full Product Details