Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HDA05
Entrez Gene ID
836227
Gene Name
histone deacetylase 5
Gene Aliases
AT5G61060, ATHDA5, HDA5, MAF19.7, MAF19_7, histone deacetylase 5
Chromosome Location
Chr.5: 24566783 - 24571072 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001203654.1 NP_001190583.1 7 - 8 1200 73
NM_125499.5 NP_200914.2 7 - 8 1269 73
GenBank mRNA AY090936.1 - 7 - 8 1188 73
AY120784.1 - 7 - 8 1182 73
BT000073.1 - 7 - 8 1114 73

View Full Product Details