Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- CYP89A3
- Entrez Gene ID
-
836253
- Gene Name
- cytochrome P450, family 89, subfamily A, polypeptide 3
- Gene Aliases
-
''cytochrome P450, AT5G61320, MFB13.19, MFB13_19, cytochrome P450, family 89, polypeptide 3, polypeptide 3'', subfamily A
- Chromosome Location
-
Chr.5: 24655098 - 24656638 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_125525.1 |
NP_200940.1 |
1 - 2 |
960 |
121 |
View Full Product Details