Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- CYP81K2
- Entrez Gene ID
-
830925
- Gene Name
- cytochrome P450, family 81, subfamily K, polypeptide 2
- Gene Aliases
-
''cytochrome P450, AT5G10600, F12B17.50, F12B17_50, cytochrome P450, family 81, polypeptide 2, polypeptide 2'', subfamily K
- Chromosome Location
-
Chr.5: 3351037 - 3352894 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_121098.3 |
NP_196622.2 |
1 - 1 |
728 |
84 |
- Gene Symbol
- AT5G10605
- Entrez Gene ID
-
10723039
- Gene Name
- methyltransferase
- Gene Aliases
-
AT5G10605
- Chromosome Location
-
Chr.5: 3351196 - 3353392 on Build TAIR10.1
View Full Product Details