Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
EMF1
Entrez Gene ID
831025
Gene Name
embryonic flower 1 (EMF1)
Gene Aliases
AT5G11530, F15N18.120, F15N18_120, embryonic flower 1
Chromosome Location
Chr.5: 3695792 - 3701549 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001343200.1 NP_001330242.1 4 - 5 1091 149
NM_001343201.1 NP_001330243.1 4 - 5 813 149
NM_121191.4 NP_196714.1 4 - 5 844 149

View Full Product Details