Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cwINV6
Entrez Gene ID
831065
Gene Name
6-&1-fructan exohydrolase
Gene Aliases
6-&1-fructan exohydrolase, AT5G11920, AtcwINV6, F14F18.90, F14F18_90
Chromosome Location
Chr.5: 3839316 - 3842409 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001085105.1 NP_001078574.1 4 - 5 1230 107
NM_001343222.1 NP_001330604.1 3 - 4 1575 107
NM_121230.3 NP_568254.1 4 - 5 1305 107
GenBank mRNA AY060553.1 - 4 - 5 1230 107
AY141991.1 - 4 - 5 1131 107

View Full Product Details