Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DBP1
Entrez Gene ID
817102
Gene Name
DNA-binding protein phosphatase 1
Gene Aliases
AT2G25620, AtDBP1, DNA-binding protein phosphatase 1
Chromosome Location
Chr.2: 10902716 - 10905287 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001336005.1 NP_001324148.1 3 - 4 893 77
NM_128120.2 NP_180133.1 3 - 4 913 77
GenBank mRNA AF326901.1 - 3 - 4 871 77
AF339719.1 - 3 - 4 604 77
AF436827.1 - 3 - 4 875 77
AY039595.1 - 3 - 4 871 77
AY057740.1 - 3 - 4 604 77
BT002487.1 - 3 - 4 875 77

View Full Product Details