Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SPT42
Entrez Gene ID
836487
Gene Name
SPT4 homolog 2
Gene Aliases
AT5G63670, MBK5.15, MBK5_15, SPT4 homolog 2, SPT4-2
Chromosome Location
Chr.5: 25487927 - 25489600 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001203680.1 NP_001190609.1 4 - 5 376 88
NM_125762.4 NP_568976.1 4 - 5 362 88
GenBank mRNA AY035161.1 - 4 - 5 327 88
AY063032.1 - 4 - 5 296 88
AY084523.1 - 4 - 5 325 88

View Full Product Details