Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MAF4
Entrez Gene ID
836631
Gene Name
K-box region and MADS-box transcription factor family protein
Gene Aliases
AGAMOUS-like 69, AGL69, AT5G65070, F15O5.3, F15O5_3, FCL4, MADS AFFECTING FLOWERING 4
Chromosome Location
Chr.5: 25992193 - 25996202 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001126027.1 NP_001119499.1 7 - 8 641 74
NM_001203688.1 NP_001190617.1 7 - 8 661 74
NM_001345664.1 NP_001332255.1 4 - 5 940 74
NM_001345665.1 NP_001332256.1 6 - 7 840 74
NM_001345666.1 NP_001332257.1 5 - 6 452 74
NM_125906.4 NP_201312.2 6 - 7 615 74
GenBank mRNA AY231450.1 - 6 - 7 548 74

View Full Product Details