Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PEPKR2
Entrez Gene ID
837824
Gene Name
phosphoenolpyruvate carboxylase-related kinase 2
Gene Aliases
AT1G12680, T12C24.32, T12C24_32, phosphoenolpyruvate carboxylase-related kinase 2
Chromosome Location
Chr.1: 4319877 - 4323265 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_101138.4 NP_172728.1 3 - 4 1650 73
GenBank mRNA AY062754.1 - 3 - 4 1330 73
BT006608.1 - 2 - 3 1047 73

View Full Product Details