Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MAP65-7
Entrez Gene ID
838034
Gene Name
microtubule-associated protein 65-7
Gene Aliases
AT1G14690, F10B6.8, F10B6_8, microtubule-associated protein 65-7
Chromosome Location
Chr.1: 5051656 - 5055299 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001084068.3 NP_001077537.1 6 - 7 1204 94
NM_101338.4 NP_172922.2 5 - 6 1287 94
GenBank mRNA AY120768.1 - 5 - 6 1222 94
BT008373.1 - 5 - 6 924 94

View Full Product Details