Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
AUL1
Entrez Gene ID
843868
Gene Name
auxin-like 1 protein
Gene Aliases
AT1G75310, auxilin-like 1, auxin-like 1 protein
Chromosome Location
Chr.1: 28260801 - 28266323 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001334682.1 NP_001320612.1 6 - 7 4320 81
NM_106186.2 NP_177666.1 6 - 7 4341 81

View Full Product Details