Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LAC7
Entrez Gene ID
820078
Gene Name
laccase 7
Gene Aliases
AT3G09220, laccase 7
Chromosome Location
Chr.3: 2827190 - 2830655 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001337807.1 NP_001326525.1 2 - 3 1041 81
NM_111756.3 NP_187533.1 3 - 4 666 81
GenBank mRNA AK117639.1 - 3 - 4 553 81
BT004971.1 - 3 - 4 488 81

View Full Product Details