Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ATPK7
Entrez Gene ID
822380
Gene Name
Protein kinase superfamily protein
Gene Aliases
AT3G27580, D6 PROTEIN KINASE LIKE 3, D6PKL3
Chromosome Location
Chr.3: 10217417 - 10220061 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001035707.3 NP_001030784.1 2 - 3 1199 101
NM_113674.4 NP_189395.1 1 - 2 1430 101
GenBank mRNA AK175529.1 - 2 - 3 1038 101
AK176399.1 - 1 - 2 1345 101
BT010571.1 - 1 - 2 853 101

View Full Product Details