Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RCK
Entrez Gene ID
822395
Gene Name
DNA helicase ROCK-N-ROLLERS
Gene Aliases
AT3G27730, MER3, ROCK-N-ROLLERS
Chromosome Location
Chr.3: 10273098 - 10280646 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001338905.1 NP_001326065.1 15 - 16 1738 106
NM_001338906.1 NP_001326062.1 15 - 16 1648 106
NM_001338907.1 NP_001326064.1 15 - 16 1585 106
NM_001338908.1 NP_001326061.1 14 - 15 1611 106
NM_001338909.1 NP_001326060.1 15 - 16 1739 106
NM_001338910.1 NP_001326063.1 15 - 16 1600 106
NM_113689.2 NP_189410.2 14 - 15 1611 106
GenBank mRNA AY822649.1 - 14 - 15 1462 106
AY960701.1 - 14 - 15 1611 106

View Full Product Details