Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ABCA6
Entrez Gene ID
823931
Gene Name
ABC2 homolog 5
Gene Aliases
A. THALIANA ABC2 HOMOLOG 5, ABC2 homolog 5, AT3G47770, ATATH5, ATH5, ATP-binding cassette A6
Chromosome Location
Chr.3: 17617972 - 17622915 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001339335.1 NP_001327229.1 1 - 2 151 71
NM_001339336.1 NP_001327230.1 1 - 2 151 71
NM_114645.3 NP_190361.2 1 - 2 68 71

View Full Product Details