Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PLDALPHA3
Entrez Gene ID
832609
Gene Name
phospholipase D alpha 3
Gene Aliases
AT5G25370, F18G18.110, F18G18_110, phospholipase D alpha 3
Chromosome Location
Chr.5: 8803829 - 8807783 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001343920.1 NP_001318645.1 2 - 3 155 147
NM_001343921.1 NP_001331647.1 2 - 3 212 147
NM_122446.2 NP_197919.1 1 - 2 325 147

View Full Product Details