Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ASD2
Entrez Gene ID
832681
Gene Name
alpha-L-arabinofuranosidase 2
Gene Aliases
ARABIDOPSIS THALIANA ALPHA-L-ARABINOFURANOSIDASE 2, AT5G26120, ATASD2, T1N24.13, T1N24_13, alpha-L-arabinofuranosidase 2
Chromosome Location
Chr.5: 9121675 - 9125967 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_122513.4 NP_197984.2 12 - 13 1379 91
GenBank mRNA AY063925.1 - 12 - 13 1379 91
AY243510.1 - 11 - 12 1309 91
BT006062.1 - 11 - 12 1304 91

View Full Product Details