Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- ATK4
- Entrez Gene ID
-
832758
- Gene Name
- kinesin 4
- Gene Aliases
-
AT5G27000, F2P16.12, F2P16_12, KATD, KINESIN-LIKE PROTEIN IN ARABIDOPSIS THALIANA D, kinesin 4
- Chromosome Location
-
Chr.5: 9497939 - 9503173 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_122582.2 |
NP_568491.1 |
11 - 12 |
1623 |
61 |
View Full Product Details