Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ALB4
Entrez Gene ID
839065
Gene Name
OxaA/YidC-like membrane insertion protein
Gene Aliases
ALBINA 4, ARABIDOPSIS THALIANA ENVELOPE MEMBRANE INTEGRASE, ARTEMIS, AT1G24490, F21J9.16
Chromosome Location
Chr.1: 8682195 - 8685271 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001160892.1 NP_001154364.1 3 - 4 597 104
NM_102294.7 NP_173858.5 4 - 5 747 104
GenBank mRNA AM177312.1 - 4 - 5 708 104

View Full Product Details