Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RLP13
Entrez Gene ID
843757
Gene Name
receptor like protein 13
Gene Aliases
AT1G74170, AtRLP13, F9E11.7, F9E11_7, receptor like protein 13
Chromosome Location
Chr.1: 27891494 - 27896436 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001334634.1 NP_001322432.1 6 - 7 971 153
NM_106077.4 NP_177557.2 7 - 8 1323 153

View Full Product Details