Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ACBP5
Entrez Gene ID
832825
Gene Name
acyl-CoA binding protein 5
Gene Aliases
AT5G27630, AtACBP5, F15A18.90, F15A18_90, acyl-CoA binding protein 5
Chromosome Location
Chr.5: 9775854 - 9781002 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001344021.1 NP_001331944.1 7 - 8 996 85
NM_001344022.1 NP_001331946.1 7 - 8 996 85
NM_001344023.1 NP_001331945.1 7 - 8 827 85
NM_122645.5 NP_198115.2 7 - 8 853 85
GenBank mRNA AY093156.1 - 7 - 8 827 85
BT008399.1 - 7 - 8 749 85

View Full Product Details