Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CUE1
Entrez Gene ID
833308
Gene Name
Glucose-6-phosphate/phosphate translocator-like protein
Gene Aliases
ARABIDOPSIS THALIANA PHOSPHATE/PHOSPHOENOLPYRUVATE TRANSLOCATOR, ARAPPT, AT5G33320, CAB UNDEREXPRESSED 1, F19N2.40, F19N2_40, PHOSPHOENOLPYRUVATE/PHOSPHATE TRANSLOCATOR, PPT
Chromosome Location
Chr.5: 12588741 - 12591748 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_122856.3 NP_198317.1 8 - 9 1361 50
GenBank mRNA AY080788.1 - 8 - 9 1264 50
AY133809.1 - 8 - 9 1152 50

View Full Product Details