Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MRN1
Entrez Gene ID
834267
Gene Name
marneral synthase
Gene Aliases
AT5G42600, MFO20.1, MFO20_1, marneral synthase, marneral synthase 1
Chromosome Location
Chr.5: 17053453 - 17058245 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_123624.4 NP_199074.1 8 - 9 1349 86
GenBank mRNA BT020312.1 - 8 - 9 1285 86
BT020456.1 - 8 - 9 1236 86

View Full Product Details