Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RGP4
Entrez Gene ID
835147
Gene Name
reversibly glycosylated polypeptide 4
Gene Aliases
AT5G50750, MFB16.25, MFB16_25, reversibly glycosylated polypeptide 4
Chromosome Location
Chr.5: 20640992 - 20642787 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_124453.3 NP_199888.1 2 - 3 658 111
GenBank mRNA AF329280.1 - 2 - 3 596 111
BT004025.1 - 2 - 3 622 111
BT005194.1 - 2 - 3 584 111
KJ138643.1 - 2 - 3 584 111

View Full Product Details