Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FEI2
Entrez Gene ID
818130
Gene Name
Leucine-rich repeat protein kinase family protein
Gene Aliases
AT2G35620, FEI 2, T20F21.18, T20F21_18
Chromosome Location
Chr.2: 14960851 - 14965497 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001202755.2 NP_001189684.1 6 - 7 814 124
NM_001336565.1 NP_001324113.1 6 - 7 1007 124
NM_001336566.1 NP_001324112.1 6 - 7 753 124
NM_129116.5 NP_181105.2 6 - 7 778 124
GenBank mRNA FJ708709.1 - 5 - 6 453 124

View Full Product Details