Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LAC5
Entrez Gene ID
818630
Gene Name
laccase 5
Gene Aliases
AT2G40370, T3G21.14, T3G21_14, laccase 5
Chromosome Location
Chr.2: 16857959 - 16860779 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_129597.4 NP_181568.1 4 - 5 804 138
GenBank mRNA AF506030.1 - 4 - 5 631 138

View Full Product Details