Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DPE2
Entrez Gene ID
818682
Gene Name
disproportionating enzyme 2
Gene Aliases
AT2G40840, T20B5.4, T20B5_4, disproportionating enzyme 2
Chromosome Location
Chr.2: 17044932 - 17051062 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_129647.4 NP_181616.3 22 - 23 2959 68
GenBank mRNA AY081315.1 - 22 - 23 2750 68
BT010364.1 - 21 - 22 2701 68

View Full Product Details