Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
COS1
Entrez Gene ID
819010
Gene Name
6,7-dimethyl-8-ribityllumazine synthase / DMRL synthase / lumazine synthase / riboflavin synthase
Gene Aliases
AT2G44050, COI1 SUPPRESSOR1, F6E13.18, coronatine insensitive1 suppressor
Chromosome Location
Chr.2: 18224213 - 18226233 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_129967.4 NP_181933.1 3 - 4 457 97
GenBank mRNA AF148649.1 - 3 - 4 366 97
AY050913.1 - 3 - 4 406 97
AY091343.1 - 3 - 4 366 97

View Full Product Details