Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MAP2A
Entrez Gene ID
819025
Gene Name
methionine aminopeptidase 2A
Gene Aliases
AT2G44180, F6E13.31, methionine aminopeptidase 2A
Chromosome Location
Chr.2: 18269211 - 18271941 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001337079.1 NP_001323713.1 7 - 8 1218 130
NM_129981.5 NP_566013.1 8 - 9 1150 130
GenBank mRNA AK228284.1 - 8 - 9 1053 130
BT006476.1 - 8 - 9 995 130

View Full Product Details