Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- ERD15
- Entrez Gene ID
-
818741
- Gene Name
- dehydration-induced protein (ERD15)
- Gene Aliases
-
AT2G41430, CID1, CTC-Interacting Domain 1, DEHYDRATION-INDUCED PROTEIN, EARLY RESPONSIVE TO DEHYDRATION 15, LIGHT STRESS-REGULATED 1, LSR1, T26J13.2
- Chromosome Location
-
Chr.2: 17269158 - 17270542 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_201928.2 |
NP_973657.1 |
2 - 2 |
583 |
96 |
View Full Product Details