Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PDLP4
Entrez Gene ID
819592
Gene Name
plasmodesmata-located protein 4
Gene Aliases
AT3G04370, T27C4.1, T27C4_1, plasmodesmata-located protein 4
Chromosome Location
Chr.3: 1159409 - 1161338 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001035554.2 NP_001030631.1 1 - 2 468 133

View Full Product Details