Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
CHIC2
Entrez Gene ID
615190
Gene Name
cysteine rich hydrophobic domain 2
Gene Aliases
-
Chromosome Location
Chr.6: 69536633 - 69596055 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001078048.1 NP_001071516.1 2 - 3 508 103
GenBank mRNA BC123812.1 - 2 - 3 508 103

View Full Product Details