The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_173991.2 | NP_776416.1 | 3 - 4 | 296 | 66 |
GenBank mRNA | BC102620.1 | - | 3 - 4 | 308 | 66 |
BC134729.1 | - | 3 - 4 | 276 | 66 | |
BT021053.1 | - | 3 - 4 | 295 | 66 | |
X61171.1 | - | - | 258 | 66 | |
X64839.1 | - | - | 296 | 66 |