Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PROCR
Entrez Gene ID
282005
Gene Name
protein C receptor
Gene Aliases
EPCR
Chromosome Location
Chr.13: 64444253 - 64449124 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_174437.1 NP_776862.1 3 - 4 750 68
GenBank mRNA BC116074.1 - 3 - 4 695 68
L39065.1 - 3 - 4 750 68

View Full Product Details