Assay Design

The amplified product spans an exon junction and the probe and one of the primers sit within one exon. This assay may detect gDNA.

Gene Transcripts

Gene Symbol
VSX1
Entrez Gene ID
286802
Gene Name
visual system homeobox 1
Gene Aliases
RINX
Chromosome Location
Chr.13: 42691095 - 42699689 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_174767.2 NP_777192.1 1 - 1 147 113
GenBank mRNA AF251032.1 - 1 - 1 147 113
BC133552.1 - 1 - 1 399 113

View Full Product Details