Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RRNAD1
Entrez Gene ID
532204
Gene Name
ribosomal RNA adenine dimethylase domain containing 1
Gene Aliases
C1orf66, C3H1orf66, CGI-41
Chromosome Location
Chr.3: 14098948 - 14105919 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001015641.1 NP_001015641.1 2 - 3 492 133
GenBank mRNA BC112638.1 - 2 - 3 387 133
BT020816.1 - 2 - 3 492 133

View Full Product Details