Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
AIMP1
Entrez Gene ID
505126
Gene Name
aminoacyl tRNA synthetase complex interacting multifunctional protein 1
Gene Aliases
SCYE1, p43
Chromosome Location
Chr.6: 18837531 - 18864984 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001035018.2 NP_001030190.2 3 - 4 291 151
GenBank mRNA BC103049.1 - 3 - 4 292 151

View Full Product Details