Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CRISP3
Entrez Gene ID
618612
Gene Name
cysteine-rich secretory protein 3
Gene Aliases
-
Chromosome Location
Chr.23: 22447442 - 22472520 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001079635.1 NP_001073103.1 1 - 2 104 89
GenBank mRNA BC102058.1 - 1 - 2 104 89
BT030687.1 - 1 - 2 113 89

View Full Product Details