Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KIR2DL5A
Entrez Gene ID
282860
Gene Name
killer cell immunoglobulin-like receptor, two domains, long cytoplasmic tail, 5A
Gene Aliases
KIR2DL1, KIR2DL4
Chromosome Location
Chr.18: 62542299 - 62552522 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_174740.2 NP_777165.1 2 - 3 75 75
GenBank mRNA AF490399.1 - 2 - 3 75 75
AY075102.1 - 2 - 3 75 75

View Full Product Details