Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FDPS
Entrez Gene ID
281156
Gene Name
farnesyl diphosphate synthase
Gene Aliases
-
Chromosome Location
Chr.3: 15311071 - 15320374 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_177497.2 NP_803463.1 7 - 8 737 101
GenBank mRNA AF461050.1 - 7 - 8 737 101
BC111256.1 - 8 - 9 1557 101
BC120218.1 - 8 - 9 799 101
BC149572.1 - 8 - 9 858 101

View Full Product Details