Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PSMB6
Entrez Gene ID
510069
Gene Name
proteasome 20S subunit beta 6
Gene Aliases
-
Chromosome Location
Chr.19: 26564855 - 26567068 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034369.2 NP_001029541.1 5 - 6 586 66
GenBank mRNA BC105176.1 - 5 - 6 587 66

View Full Product Details