Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ARID5A
Entrez Gene ID
524118
Gene Name
AT-rich interaction domain 5A
Gene Aliases
-
Chromosome Location
Chr.11: 2557603 - 2569332 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001035349.1 NP_001030426.1 2 - 3 180 117
GenBank mRNA BC104604.1 - 2 - 3 180 117

View Full Product Details