The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001037452.1 | NP_001032529.1 | 6 - 7 | 483 | 91 |
GenBank mRNA | BC108165.1 | - | 6 - 7 | 483 | 91 |