Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RNPC3
Entrez Gene ID
768060
Gene Name
RNA binding region (RNP1, RRM) containing 3
Gene Aliases
-
Chromosome Location
Chr.3: 39837781 - 39883488 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001077117.2 NP_001070585.1 6 - 7 694 73
GenBank mRNA BC105165.1 - 6 - 7 695 73

View Full Product Details