Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DSB
Entrez Gene ID
618722
Gene Name
MHC class II antigen DS beta
Gene Aliases
-
Chromosome Location
Chr.23: 7035502 - 7055197 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001083785.1 NP_001077254.1 2 - 3 371 88

View Full Product Details