Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NT5C3A
Entrez Gene ID
511858
Gene Name
5'-nucleotidase, cytosolic IIIA
Gene Aliases
NT5C3
Chromosome Location
Chr.4: 63768335 - 63807506 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001037597.1 NP_001032686.1 9 - 10 889 130
GenBank mRNA BC109771.1 - 9 - 10 889 130

View Full Product Details