Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ARFRP1
Entrez Gene ID
525394
Gene Name
ADP ribosylation factor related protein 1
Gene Aliases
-
Chromosome Location
Chr.13: 54047379 - 54053623 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001037607.1 NP_001032696.1 6 - 7 476 63
GenBank mRNA BC109550.1 - 6 - 7 476 63

View Full Product Details