Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TEKT4
Entrez Gene ID
510343
Gene Name
tektin 4
Gene Aliases
-
Chromosome Location
Chr.25: 872389 - 878381 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001038069.2 NP_001033158.1 4 - 5 1078 60
GenBank mRNA BC111132.1 - 4 - 5 1079 60

View Full Product Details