Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
METTL17
Entrez Gene ID
514880
Gene Name
methyltransferase like 17
Gene Aliases
METT11D1
Chromosome Location
Chr.10: 26119572 - 26125218 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001038542.2 NP_001033631.1 13 - 14 1289 97
GenBank mRNA BC109846.1 - 13 - 14 1290 97

View Full Product Details